Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AAMDC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558164
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558164 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558164 Supplier Invitrogen™ Supplier No. PA558164
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EIASLSWGQM KVKGSNTTYK DCKVWPGGSR TWDWRETGTE HSPGVQPADV KEVVEK Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.

AAMDC (C11orf67) may play a role in preadipocyte differentiation and adipogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AAMDC
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AAMDC
Gene Accession No. Q9H7C9
Gene Alias 1810020D17Rik; 1810037D19Rik; Aamdc; adipogenesis associated Mth938 domain containing; adipogenesis associated Mth938 domain-containing protein; adipogenesis associated, Mth938 domain containing; C11orf67; C29H11orf67; CK067; hypothetical protein LOC553802; LI2; Mth938 domain-containing protein; PTD015; RGD1561459; Unknown (protein for MGC:133476); UPF0366 protein C11orf67; UPF0366 protein C11orf67 homolog; zgc:112239
Gene Symbols AAMDC
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human AAMDC. Recombinant protein control fragment (Product #RP-97281).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28971
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.