Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AANAT Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA1760

Catalog No. PA1760


Only null left
Edge
Add to Cart

Description

Description

PA1-760 detects serotonin N-acetyltransferase (AA-NAT) from rat tissues. PA1-760 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~23 kDa protein representing unphosphorylated and phosphorylated AA-NAT from rat pineal gland whole protein extract. PA1-760 immunizing phosphopeptide (Cat. # PEP-064) corresponds to amino-terminal amino acid residues 24-33 of mouse AA-NAT. Total anti-amino-terminal AA-NAT-specific IgG (Cat. # PA1-760) was affinity purified on resin-coupled non-phosphopeptide (Cat. # PEP-063). Phospho-specific amino-terminal IgG (Cat. # PA1-761) was subsequently isolated on resin-coupled phosphopeptide (Cat. # PEP-064). This sequence is completely conserved between mouse, rat, and human AA-NAT protein.

Arylalkylamine N-Acetyltransferase (AANAT) is the penultimate enzyme is melatonin synthesis. Hormonal melatonin has been implicated in sleep and circadian clock function. Large changes in the abundance of AANAT are believed to be responsible for the large day/night rhythms in melatonin synthesis in the pineal gland and retina. Most research shows that AANAT activity is regulated by controlling the steady-state levels of the protein. Recent research, however, suggests that another mechanism may also play a role in controlling melatonin production without altering AANAT protein and activity. This mechanism may explain the sudden physiological increase in circulating melatonin observed at the onset of the dark period in humans.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

AANAT
Polyclonal
Unconjugated
Aanat
Aanat; AA-NAT; Aralkylamine N-acetyltransferase; arylakylamine N-acetyltransferase; Arylalkylamine N - acetyltransferase (Serotonin N - acetyltransferase); arylalkylamine N-acetyltransferase; DSPS; Nat-2; Nat4; Seretonin N-acetyltransferase; serotonin acetylase; Serotonin N-acetyltransferase; SNAT
Rabbit
Antigen affinity chromatography
RUO
25120
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
1 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide
Q64666
Aanat
Synthetic Peptide: C(24) Q R R H T L P A S(33).
50 μg
Primary
Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.