Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AANAT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565859
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565859 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565859 Supplier Invitrogen™ Supplier No. PA565859
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FLTLCPELSLG WFEEGCLVAF IIGSLWDKER LMQESLTLHR SGGHIAHLHV LAVHRAFRQQ GRGPILLWR Highest antigen sequence identity to the following orthologs - mouse 89%, rat 89%.

Arylalkylamine N-Acetyltransferase (AANAT) is the penultimate enzyme is melatonin synthesis. Hormonal melatonin has been implicated in sleep and circadian clock function. Large changes in the abundance of AANAT are believed to be responsible for the large day/night rhythms in melatonin synthesis in the pineal gland and retina. Most research shows that AANAT activity is regulated by controlling the steady-state levels of the protein. Recent research, however, suggests that another mechanism may also play a role in controlling melatonin production without altering AANAT protein and activity. This mechanism may explain the sudden physiological increase in circulating melatonin observed at the onset of the dark period in humans.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AANAT
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Aanat
Gene Accession No. Q16613
Gene Alias Aanat; AA-NAT; Aralkylamine N-acetyltransferase; arylakylamine N-acetyltransferase; Arylalkylamine N - acetyltransferase (Serotonin N - acetyltransferase); arylalkylamine N-acetyltransferase; DSPS; Nat-2; Nat4; Seretonin N-acetyltransferase; serotonin acetylase; Serotonin N-acetyltransferase; SNAT
Gene Symbols Aanat
Host Species Rabbit
Immunogen Recombinant Human Serotonin N-AT. Recombinant protein control fragment (Product #RP-106527).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.