Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AAO Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598359
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598359 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598359 Supplier Invitrogen™ Supplier No. PA598359
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AAO Polyclonal Antibody for Western Blot, ELISA

AAO is an enzyme that may be involved in a redox system involving ascorbic acid.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AAO
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.48 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Host Species Rabbit
Immunogen Recombinant Cucurbita maxima L-ascorbate oxidase protein (31-579AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Plant
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.