Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AAQ Chloride

Catalog No. 546250
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
546250 50 mg
546210 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 546250 Supplier Tocris Bioscience™ Supplier No. 5462/50
This item is not returnable. View return policy

Photoswitchable Kv channel blocker

AAQ chloride is a photoswitchable Kv channel blocker (IC50 values are 2 and 64 μM at 500 nm and 380 nm respectively). Switches conformation from cis to trans at 500 nm and trans to cis at 380 nm. Exhibits minimal activity at Nav1.2 and L-type Ca2+ channels. Stimulates action potential firing of hippocampal neurons in vitro at 500 nm and restores visual responsiveness in blind mice at 380 nm.

Specifications

Chemical Name or Material N,N,N-Triethyl-2-oxo-2-[[4-[2-[4-[(1-oxo-2-propen-1-yl)amino]phenyl]diazenyl]phenylamino]ethanaminium chloride
Quantity 50 mg
Solubility Soluble to 20mM in DMSO
Molecular Formula C23H30ClN5O2
Formula Weight Observed MW: 443.97
Purity >98%

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.