Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AARS2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$254.42 - $665.78
Specifications
| Antigen | AARS2 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
AARS2 Polyclonal specifically detects AARS2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| AARS2 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q5JTZ9 | |
| 57505 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:THLLNWALRQTLGPGTEQQGSHLNPEQLRLDVTTQTPLTPEQLRAVENTVQEAVGQDEAVYMEEVPLALTAQVP | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 107 kDa |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| alanine tRNA ligase 2, mitochondrial (putative), alanine--tRNA ligase, alanyl-tRNA synthetase 2, mitochondrial (putative), alanyl-tRNA synthetase, mitochondrial, alaRS, bA444E17.1, COXPD8, KIAA1270, MTALARS, MT-ALARS, probable alanyl-tRNA synthetase, mitochondrial | |
| AARS2 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title