Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AARS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566387
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566387 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566387 Supplier Invitrogen™ Supplier No. PA566387
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QLSNGTTRLLA VTGEQAQQAR ELGQSLAQEV KAATERLSLG SRDVAEALRL SKDIGRLIEA VETAVMP Highest antigen sequence identity to the following orthologs - mouse 75%, rat 75%.

The protein encoded by this gene belongs to the class-II aminoacyl-tRNA synthetase family. Aminoacyl-tRNA synthetases play critical roles in mRNA translation by charging tRNAs with their cognate amino acids. The encoded protein is a mitochondrial enzyme that specifically aminoacylates alanyl-tRNA. Mutations in this gene are a cause of combined oxidative phosphorylation deficiency 8. Catalyzes the attachment of alanine to tRNA(Ala) in a two-step reaction: alanine is first activated by ATP to form Ala-AMP and then transferred to the acceptor end of tRNA(Ala). Also edits incorrectly charged tRNA(Ala) via its editing domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AARS2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AARS2
Gene Accession No. Q5JTZ9
Gene Alias AARS2; AARSL; alanine tRNA ligase 2, mitochondrial (putative); alanine--tRNA ligase, mitochondrial; alanine--tRNA ligase, mitochondrial {ECO:0000255; Alanyl-tRNA synthetase; alanyl-tRNA synthetase {ECO:0000255; alanyl-tRNA synthetase 2, mitochondrial; alanyl-tRNA synthetase 2, mitochondrial (putative); alanyl-tRNA synthetase like; alanyl-tRNA synthetase, mitochondrial; alaRS; alaRS {ECO:0000255; bA444E17.1; COXPD8; Gm89; HAMAP-Rule:MF_03133}; Kiaa1270; LKENP; MTALARS; MT-ALARS; probable alanyl-tRNA synthetase, mitochondrial
Gene Symbols AARS2
Host Species Rabbit
Immunogen Recombinant Human AARS2. Recombinant protein control fragment (Product #RP-107060).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57505
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.