Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AARSD1 Antibody (012), DyLight 405, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290276V
Description
AARSD1 Monoclonal specifically detects AARSD1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
AARSD1 | |
Monoclonal | |
DyLight 405 | |
alanyl-tRNA editing protein Aarsd1, alanyl-tRNA synthetase domain containing 1, Alanyl-tRNA synthetase domain-containing protein 1, MGC2744 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human AARSD1 (Q9BTE6-2; Thr176-Glu525). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunofluorescence | |
12 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
80755 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction