Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AARSD1 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300270APC
Description
AARSD1 Polyclonal antibody specifically detects AARSD1 in Human samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
AARSD1 | |
Polyclonal | |
Western Blot, ELISA, Immunoprecipitation | |
alanyl-tRNA editing protein Aarsd1, alanyl-tRNA synthetase domain containing 1, Alanyl-tRNA synthetase domain-containing protein 1, MGC2744 | |
Produced in rabbits immunized with purified, recombinant Human AARSD1 (Uniprot#: Q9BTE6-2; Thr176-Glu525). | |
0.1 mL | |
Cancer, Cellular Signaling, Epigenetics | |
80755 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunoprecipitation | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction