Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AARSD1 Antibody (Tsp232A), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212887H
Description
AARSD1 Monoclonal specifically detects AARSD1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
AARSD1 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
AARSD1 | |
Recombinant tsp23 | |
0.1 mL | |
80755 | |
Human, Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Tsp232A | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
alanyl-tRNA editing protein Aarsd1, alanyl-tRNA synthetase domain containing 1, Alanyl-tRNA synthetase domain-containing protein 1, MGC2744 | |
Mouse | |
Protein G purified | |
Primary | |
Detects approx 19kDa. Detects Tsp23, no cross-reactivity to p23. | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction