Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AARSD1 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA519203
Description
This antibody is predicted to react with bovine and canine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
AARSD1 functions in trans to edit the amino acid moiety from incorrectly charged tRNA(Ala).
Specifications
AARSD1 | |
Polyclonal | |
Unconjugated | |
AARSD1 | |
1110069E20Rik; 2310044P18Rik; AA589600; AARSD1; Alanyl-tRNA deacylase alaX; alanyl-tRNA editing protein Aarsd1; alanyl-tRNA synthetase domain containing 1; alanyl-tRNA synthetase domain-containing protein 1; Alax; AlaXp-II; MGC2744 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
80755 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9BTE6 | |
AARSD1 | |
Synthetic peptide sequence (IEFYAKVNSKDSQDK) corresponding to the internal amino acids of AARSD1 (aa 54-68). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction