Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PTGES3L-AARSD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554399
Description
Immunogen sequence: LKKVSTKRPP PAMDDLDFTT TVVSCCPAEL QTEGSNGKKE VLSGFQVVLE DTVLFPEGGG QPDDRGTIND ISVLRVTRRG EQADHFTQTP LDPGSQVLVR V Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 75%.
This locus represents naturally occurring readthrough transcription between the neighboring PTGES3L (prostaglandin E synthase 3 (cytosolic)-like) and AARSD1(alanyl-tRNA synthetase domain containing 1) genes on chromosome 17. The readthrough transcript encodes a fusion protein that shares sequence identity with each individual gene product. [provided by RefSeq, May 2012].
Specifications
PTGES3L-AARSD1 | |
Polyclonal | |
Unconjugated | |
PTGES3L-AARSD1 | |
AARSD1; Alanyl-TRNA Synthetase Domain-Containing Protein 1; PTGES3L | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100885850 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BTE6 | |
PTGES3L-AARSD1 | |
Recombinant protein corresponding to Human AARSD1. Recombinant protein control fragment (Product #RP-93207). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction