Learn More
Invitrogen™ AARSD1 Recombinant Rabbit Monoclonal Antibody (012)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA528987
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human AARSD1.
AARSD1 functions in trans to edit the amino acid moiety from incorrectly charged tRNA(Ala).
Specifications
AARSD1 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q9BTE6 | |
AARSD1 | |
Recombinant Human AARSD1 Protein. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Western Blot, Immunocytochemistry | |
012 | |
Unconjugated | |
AARSD1 | |
1110069E20Rik; 2310044P18Rik; AA589600; AARSD1; Alanyl-tRNA deacylase alaX; alanyl-tRNA editing protein Aarsd1; alanyl-tRNA synthetase domain containing 1; alanyl-tRNA synthetase domain-containing protein 1; Alax; AlaXp-II; MGC2744 | |
Rabbit | |
Protein A | |
RUO | |
80755 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.