Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AASD-PPT Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP189322
Description
AASD-PPT Polyclonal specifically detects AASD-PPT in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
AASD-PPT | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Q9NRN7 | |
AASDHPPT | |
This antibody was developed against Recombinant Protein corresponding to amino acids:EWAFEESKIDEHHFVAVALRKPDGSRHQDVPSQDDSKPTQRQFTILNFNDLMSSAVPMTPEDPSFWDCFCFTEEIPIRNGTKS | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
4'-phosphopantetheinyl transferase, AASD-PPTDKFZp566E2346, Alpha-aminoadipic semialdehyde dehydrogenase-phosphopantetheinyl transferase, aminoadipate-semialdehyde dehydrogenase-phosphopantetheinyl transferase, CGI-80, EC 2.7.8, EC 2.7.8.-, L-aminoadipate-semialdehyde dehydrogenase-phosphopantetheinyl transferase, LYS2, LYS5, LYS5 ortholog | |
Rabbit | |
36 kDa | |
0.1 mL | |
Proteases & Other Enzymes | |
60496 | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction