Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AASDH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557675
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557675 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557675 Supplier Invitrogen™ Supplier No. PA557675
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RKLSDINQEE ASGTSLHQKA IMTFTCHNEI NAFVVLSRGS QILSLNSTRF LTKLGHCSSA CPSDSVSQTN IQNLKGLNSP VLIGKSKDPS CV Highest antigen sequence identity to the following orthologs: Mouse - 39%, Rat - 40%.

AASDH encodes a member of the non-ribosome peptide syntesase (NRPS) enzyme family. The encoded protein contains an AMP-binding domain, PP-binding (phosphopantetheine, or pantetheine 4'phosphate-binding) domain and the Pyrrolo-quinoline quinon (PQQ) binding domain. The protein is expressed in several adult tissues. Covalently binds beta-alanine in an ATP-dependent manner to form a thioester bond with its phosphopantetheine group and transfers it to an, as yet, unknown acceptor. May be required for a post-translational protein modification or for post-transcriptional modification of an RNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AASDH
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AASDH
Gene Accession No. Q4L235
Gene Alias 2-aminoadipic 6-semialdehyde dehydrogenase; 2-aminoadipic 6-semialdehyde dehydrogenase antibody; A230062G08Rik; A830035E16; Aasdh; AASDH antibody; ACSF4; ACSF4 antibody; acyl-CoA synthetase family member 4; acyl-CoA synthetase family member 4 antibody; aminoadipate-semialdehyde dehydrogenase; aminoadipate-semialdehyde dehydrogenase antibody; Beta-alanine-activating enzyme; HSPC318; LYS2; LYS2 antibody; non-ribosomal peptide synthetase 1098; non-ribosomal peptide synthetase 1098 antibody; non-ribosomal peptide synthetase 998; non-ribosomal peptide synthetase 998 antibody; NRPS1098; NRPS1098 antibody; NRPS998; NRPS998 antibody; Protein LYS2 homolog; Protein NRPS998; putative aminoadipate-semialdehyde dehydrogenase; RGD1311135; U26
Gene Symbols AASDH
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human AASDH. Recombinant protein control fragment (Product #RP-96698).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 132949
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.