Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AASDHPPT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101646
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101646 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101646 Supplier Invitrogen™ Supplier No. PA5101646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total AASDHPPT.

The protein encoded by this gene is similar to Saccharomyces cerevisiae LYS5, which is required for the activation of the alpha-aminoadipate dehydrogenase in the biosynthetic pathway of lysine. Yeast alpha-aminoadipate dehydrogenase converts alpha-biosynthetic-aminoadipate semialdehyde to alpha-aminoadipate. It has been suggested that defects in the human gene result in pipecolic acidemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AASDHPPT
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene AASDHPPT
Gene Accession No. B2RYJ4, Q9CQF6, Q9NRN7
Gene Alias 2010309J24Rik; 2810407B07Rik; 4'-phosphopantetheinyl transferase; AASDHPPT; AASD-PPT; ACPS; Alpha-aminoadipic semialdehyde dehydrogenase-phosphopantetheinyl transferase; aminoadipate-semialdehyde dehydrogenase-phosphopantetheinyl transferase; CGI-80; DKFZp566E2346; HAH-P; holo ACP synthase; holo-[acyl-carrier-protein] synthase; HSPC223; L-aminoadipate-semialdehyde dehydrogenase-phosphopantetheinyl transferase; LYS2; LYS5; LYS5 ortholog; Unknown (protein for IMAGE:7962884); x0005
Gene Symbols AASDHPPT
Host Species Rabbit
Immunogen A synthesized peptide derived from human AASDHPPT(Accession Q9NRN7), corresponding to amino acid residues P264-S309.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 300328, 60496, 67618
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.