Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AASS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545462
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545462 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545462 Supplier Invitrogen™ Supplier No. PA545462
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: HHHLVRKTDA VYDPAEYDKH PERYISRFNT DIAPYTTCLI NGIYWEQNTP Sequence homology: Cow: 86%; Dog: 86%; Guinea Pig: 86%; Horse: 79%; Human: 100%; Rabbit: 86%; Rat: 86%.

AASS is a bifunctional enzyme that catalyzes the first two steps in the mammalian lysine degradation pathway. The N-terminal and the C-terminal portions of this enzyme contain lysine-ketoglutarate reductase and saccharopine dehydrogenase activity, respectively, resulting in the conversion of lysine to alpha-aminoadipic semialdehyde. Mutations in the gene encoding this protein are associated with familial hyperlysinemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AASS
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene AASS
Gene Accession No. Q9UDR5
Gene Alias AASS; aass {ECO:0000312; alpha-aminoadipate semialdehyde synthase; alpha-aminoadipic semialdehyde synthase, mitochondrial; aminoadipate-semialdehyde synthase; aminoadipic semialdehyde synthase; EMBL:AAI28772.1}; LKR; LKR/SDH; LKRSDH; LOR; LOR/SDH; Lorsdh; Lysine ketoglutarate reductase; lysine oxoglutarate reductase; lysine-2-oxoglutarate reductase; lysine-ketoglutarate reductase /saccharopine dehydrogenase; saccharopine dehydrogenase; SDH
Gene Symbols AASS
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human AASS (aa 268-317).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10157
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.