Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AASS Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554260
Description
Immunogen sequence: PVSMDICKQE EKLGFLVAKQ DLVISLLPYV LHPLVAKACI TNKVNMVTAS YITPALKELE KSVEDAGITI IGELGLDPGL DHMLAMETID KAKEVGATIE SYISYCGGLP APEHSNNPLR YK Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 83%.
AASS is a bifunctional enzyme that catalyzes the first two steps in the mammalian lysine degradation pathway. The N-terminal and the C-terminal portions of this enzyme contain lysine-ketoglutarate reductase and saccharopine dehydrogenase activity, respectively, resulting in the conversion of lysine to alpha-aminoadipic semialdehyde. Mutations in the gene encoding this protein are associated with familial hyperlysinemia.
Specifications
| AASS | |
| Polyclonal | |
| Unconjugated | |
| AASS | |
| AASS; aass {ECO:0000312; alpha-aminoadipate semialdehyde synthase; alpha-aminoadipic semialdehyde synthase, mitochondrial; aminoadipate-semialdehyde synthase; aminoadipic semialdehyde synthase; EMBL:AAI28772.1}; LKR; LKR/SDH; LKRSDH; LOR; LOR/SDH; Lorsdh; Lysine ketoglutarate reductase; lysine oxoglutarate reductase; lysine-2-oxoglutarate reductase; lysine-ketoglutarate reductase /saccharopine dehydrogenase; saccharopine dehydrogenase; SDH | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10157 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UDR5 | |
| AASS | |
| Recombinant protein corresponding to Human AASS. Recombinant protein control fragment (Product #RP-92604). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction