Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AATF / Che-1 Monoclonal Antibody (1B2D8), Active Motif™
SDP

Catalog No. 501993689
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3689 100 μg
50-199-3690 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3689 Supplier Active Motif Supplier No. 61789
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The protein encoded by this gene was identified on the basis of its interaction with MAP3K12/DLK, a protein kinase known to be involved in the induction of cell apoptosis. This gene product contains a leucine zipper, which is a characteristic motif of transcription factors, and was shown to exhibit strong transactivation activity when fused to Gal4 DNA binding domain. Overexpression of this gene interfered with MAP3K12 induced apoptosis.

Specifications

Antigen AATF / Che-1
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 1B2D8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
Gene Aatf
Gene Accession No. Q9JKX4, Q9NY61
Gene Alias 4933415H02Rik; 5830465M17Rik; Aatf; apoptosis antagonizing transcription factor; Apoptosis-antagonizing transcription factor; BFR2; CHE1; Che-1; DED; HSPC277; Protein AATF; Rb-binding protein Che-1; Traube protein; Trb
Gene Symbols Aatf
Host Species Rat
Immunogen This antibody was raised against peptide comprising amino acids 1-39 of mouse AATF (Apoptosis-antagonizing transcription factor) (NP_062790).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26574, 56321
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.