Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AATF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520006
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520006 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520006 Supplier Thermo Scientific Supplier No. PA520006
Add to Cart
Add to Cart

AATF Polyclonal Antibody

AATF Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen AATF
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Aatf
Gene Accession No. Q9JKX4, Q9NY61
Gene Alias 4933415H02Rik; 5830465M17Rik; Aatf; apoptosis antagonizing transcription factor; apoptosis-antagonizing transcription factor; BFR2; CHE1; CHE-1; DED; HSPC277; Protein AATF; rb-binding protein Che-1; Traube protein; Trb
Gene Symbols Aatf
Host Species Rabbit
Immunogen a 12 amino acid peptide from near the carboxy terminus of human AATF.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26574, 56321
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.