Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AATF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106536
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106536 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106536 Supplier Invitrogen™ Supplier No. PA5106536
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total AATF.

The protein encoded by this gene was identified on the basis of its interaction with MAP3K12/DLK, a protein kinase known to be involved in the induction of cell apoptosis. This gene product contains a leucine zipper, which is a characteristic motif of transcription factors, and was shown to exhibit strong transactivation activity when fused to Gal4 DNA binding domain. Overexpression of this gene interfered with MAP3K12 induced apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AATF
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Aatf
Gene Accession No. Q9JKX4, Q9NY61, Q9QYW0
Gene Alias 4933415H02Rik; 5830465M17Rik; Aatf; apoptosis antagonizing transcription factor; apoptosis-antagonizing transcription factor; BFR2; CHE1; CHE-1; DED; HSPC277; Protein AATF; rb-binding protein Che-1; Traube protein; Trb
Gene Symbols Aatf
Host Species Rabbit
Immunogen A synthesized peptide derived from human AATF(Accession Q9NY61), corresponding to amino acid residues M1-G46.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114512, 26574, 56321
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.