Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AATK Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552646
Description
Immunogen sequence: TSGIFTDTSS DGLQARRPDV VPAFRSLQKQ VGTPDSLDSL DIPSSASDGG YEVFSPSATG PSGGQPRALD SGYDTENYES PEFVLKEAQE GCEPQAFAEL ASEGEGPGPE TRLSTSLSGL NEKNPYRDSA Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 76%.
The protein encoded by this gene contains a tyrosine kinase domain at the N-terminus and a proline-rich domain at the C-terminus. This gene is induced during apoptosis, and expression of this gene may be a necessary pre-requisite for the induction of growth arrest and/or apoptosis of myeloid precursor cells. This gene has been shown to produce neuronal differentiation in a neuroblastoma cell line.
Specifications
AATK | |
Polyclonal | |
Unconjugated | |
AATK | |
Aatk; AATYK; aatyk1; apoptosis associated tyrosine kinase; apoptosis-associated tyrosine kinase; apoptosis-associated tyrosine kinase 1B; brain apoptosis-associated tyrosine kinase; CDK5-binding protein; KIAA0641; Lemur tyrosine kinase 1; LMR1; LMTK1; mKIAA0641; p35-binding protein; p35BP; PPP1R77; protein phosphatase 1, regulatory subunit 77; serine/threonine-protein kinase LMTK1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9625 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6ZMQ8 | |
AATK | |
Recombinant protein corresponding to Human AATK. Recombinant protein control fragment (Product #RP-89815). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction