Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AATK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104417
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104417 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104417 Supplier Invitrogen™ Supplier No. PA5104417
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total LMTK1.

The protein encoded by this gene contains a tyrosine kinase domain at the N-terminus and a proline-rich domain at the C-terminus. This gene is induced during apoptosis, and expression of this gene may be a necessary pre-requisite for the induction of growth arrest and/or apoptosis of myeloid precursor cells. This gene has been shown to produce neuronal differentiation in a neuroblastoma cell line.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AATK
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene AATK
Gene Accession No. Q6ZMQ8, Q80YE4
Gene Alias Aatk; AATYK; aatyk1; apoptosis associated tyrosine kinase; apoptosis-associated tyrosine kinase; apoptosis-associated tyrosine kinase 1B; brain apoptosis-associated tyrosine kinase; CDK5-binding protein; KIAA0641; Lemur tyrosine kinase 1; LMR1; LMTK1; mKIAA0641; p35-binding protein; p35BP; PPP1R77; protein phosphatase 1, regulatory subunit 77; serine/threonine-protein kinase LMTK1
Gene Symbols AATK
Host Species Rabbit
Immunogen A synthesized peptide derived from human AATK(Accession Q6ZMQ8), corresponding to amino acid residues S523-A573.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11302, 9625
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.