Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AATK/Serine/threonine-protein kinase LMTK1 Antibody (456006), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4866J
Description
AATK/Serine/threonine-protein kinase LMTK1 Monoclonal antibody specifically detects AATK/Serine/threonine-protein kinase LMTK1 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOFSpecifications
AATK/Serine/threonine-protein kinase LMTK1 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry (Intracellular Staining), CyTOF | |
456006 | |
Intracellular Staining by Flow Cytometry, CyTOF-ready | |
AATYKp35-binding protein, apoptosis-associated tyrosine kinaseCDK5-binding protein, Brain apoptosis-associated tyrosine kinase, EC 2.7.11, KIAA0641p35BP, Lemur tyrosine kinase 1, LMR1EC 2.7.11.1, LMTK1AATYK1, serine/threonine-protein kinase LMTK1 | |
E. coli-derived recombinant human AATK, Asn421-Ala589, Accession # AAH47378 | |
0.1 mL | |
Protein Kinase | |
9625 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction