Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AATK/Serine/threonine-protein kinase LMTK1 Antibody (456006), PerCP, Novus Biologicals™
SDP

Catalog No. NB368878 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB368878 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB368878 Supplier Novus Biologicals Supplier No. FAB4866C
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AATK/Serine/threonine-protein kinase LMTK1 Monoclonal antibody specifically detects AATK/Serine/threonine-protein kinase LMTK1 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOF

Specifications

Antigen AATK/Serine/threonine-protein kinase LMTK1
Applications Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone 456006
Conjugate PerCP
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias AATYKp35-binding protein, apoptosis-associated tyrosine kinaseCDK5-binding protein, Brain apoptosis-associated tyrosine kinase, EC 2.7.11, KIAA0641p35BP, Lemur tyrosine kinase 1, LMR1EC 2.7.11.1, LMTK1AATYK1, serine/threonine-protein kinase LMTK1
Host Species Mouse
Immunogen E. coli-derived recombinant human AATK, Asn421-Ala589, Accession # AAH47378
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 9625
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.