Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ AAV-MAX Transfection Kit

Catalog No. p-9510014
Encompass_Preferred
Click to view available options
Quantity:
1 kit for 1 L of production
1 kit for 10 L of production

The AAV-MAX Transfection Kit is an essential component of the AAV-MAX Helper-Free AAV Production System. It is formulated for the transient transfection of Viral Production Cells 2.0.

The Gibco AAV-MAX Transfection Kit is an essential component of the AAV-MAX Helper-Free AAV Production System. It is formulated for the transient transfection of Viral Production Cells 2.0.

The kit includes a cationic lipid-based transfection reagent, as well as a transfection booster and production enhancer, which are designed to work together to produce adeno-associated virus (AAV) particles from Viral Production Cells 2.0 when cultured in Viral Production Medium.

Features of the AAV-MAX Transfection Kit include:

  • Components work synergistically within the AAV-MAX system to produce high AAV titers greater than 5 x 1010 vg/mL (unpurified)
  • AAV-MAX transfection reagent—cationic lipid-based transfection reagent for helper virus-free triple transfection of high-density cultures
  • AAV-MAX booster—chemically defined component that works with the transfection reagent to achieve high transfection efficiency
  • AAV-MAX enhancer—chemically defined component that enhances the production of AAV particles
  • Robust transfection protocols are adaptable to large-scale transfection
TRUSTED_SUSTAINABILITY

Specifications

Cell Line HEK293
Content And Storage • 6 mL AAV-MAX Trasfection Reagent
• 3 mL AAV-MAX Transfection Booster
• 10 mL AAV-MAX Enhancer

Reagents should be stored at 2–8°C. Do not freeze.
Culture Type Suspension Cell Culture
Description Serum-free, animal origin-free, chemically defined
Cell Type Established Cell Lines
Format Bottle
Sample Type Plasmid DNA
Transfection Technique Lipid-based Transfection
Expression System Mammalian
For Use With (Equipment) AAV-MAX System
For Use With (Application) AAV Production
Includes 10 mL Gibco AAV-MAX Enhancer, 6 mL Gibco AAV-MAX Transfection Reagent, 3 mL Gibco AAV-MAX Supplement
Product Line AAV-MAX
Product Type AAV production transfection Kit
Quantity 1 kit for 1 L of production
Reagent Type Transfection kit
Shipping Condition Wet Ice
Sterility Sterile
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.