Learn More
Invitrogen™ AAV4 Virus Polyclonal Antibody

Description
PA1-23358 detects Adeno-Associated 4 Virus in Virus samples.
Specifications
Specifications
| Antigen | AAV4 Virus |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene Alias | AAV; AAV-4; AdV4; AV4; HADV-4; Lentivirus |
| Host Species | Rabbit |
| Immunogen | Anti-Adeno-Associated Virus (AAV) polyclonal antiserum. |
| Purification Method | Affinity Chromatography |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.