Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AAV4 Virus VP3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA14107
Description
PA1-4107 detects adeno-associated 4 virus. PA1-4107 has been successfully used in Western blot procedures. The PA1-4107 immunogen is a synthetic peptide corresponding to residues N(456) F T K L R P T N F S N C(467) of the capsid VP3 protein of AAV 4.
Adeno-associated virus (AAV) is a small virus which infects humans and some other primate species. AAV is not currently known to cause disease. The virus causes a very mild immune response, lending further support to its apparent lack of pathogenicity.
Specifications
AAV4 Virus VP3 | |
Polyclonal | |
Unconjugated | |
AAV; AAV-4 VP3; AdV4; AV4; HADV-4; Lentivirus | |
Synthetic peptide corresponding to residues N(456) F T K L R P T N F S N C(467) of the capsid VP3 protein of AAV 4. | |
100 μg | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Rabbit | |
Affinity chromatography | |
RUO | |
Virus | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction