Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AAV4 VP3 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $426.00
Specifications
Antigen | AAV4 VP3 |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:1000 |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017925
![]() |
Novus Biologicals
NB10093576SS |
0.025 mg |
Each for $206.50
|
|
|||||
NB10093576
![]() |
Novus Biologicals
NB10093576 |
0.1 mg |
Each for $426.00
|
|
|||||
Description
AAV4 VP3 Polyclonal specifically detects AAV4 VP3 in Non-species specific, Virus samples. It is validated for Western Blot.Specifications
AAV4 VP3 | |
Western Blot 1:500-1:1000 | |
Polyclonal | |
Rabbit | |
Non-species specific, Virus | |
O41855 | |
Directed against a synthetic peptide within residues 450-475 of 81 kDa capsid VP3 protein of AAV 4. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
1 mg/mL | |
Western Blot | |
Unconjugated | |
RUO | |
PBS with 0.02% Sodium Azide | |
Adeno-Associated Virus, Adeno-Associated Virus (AAV), Adenovirus 4, AdV 4 | |
IgG | |
Affinity Purified | |
Adeno-Associated Virus (AAV) |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title