Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AAV5 VP3 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10093577MFV610
Description
AAV5 VP3 Polyclonal antibody specifically detects AAV5 VP3 in Nonspecies specific, Virus samples. It is validated for Western Blot, Immunohistochemistry, In-situ HybridizationSpecifications
| AAV5 VP3 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, In Situ Hybridization (ISH) | |
| Adeno-Associated Virus, Adeno-Associated Virus (AAV) | |
| Synthetic peptide within residues 525-550 of 80 kDa capsid VP3 protein of AAV5. | |
| 0.1 mL | |
| Gene Therapy, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
| Non-species specific, Virus | |
| Purified |
| Western Blot, Immunohistochemistry, In Situ Hybridization (ISH) | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction