Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABC2 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10074598V
Description
ABC2 Polyclonal antibody specifically detects ABC2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
ABC2 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
ABC2, Amplified in breast cancer gene 2 protein, chromosome 17 open reading frame 71, FLJ10587, protein SMG8, Protein smg-8 homolog, SMG8 | |
The immunogen recognized by this antibody maps to a region between residue 250 and 300 of human amplified in breast cancer gene 2 protein using the numbering given in entry NP_060619.4 (GeneID 55181). | |
0.1 mL | |
Cancer | |
55181 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction