Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABC2 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS6515R

Description
The function of this protein remains unknown.Specifications
ABC2 | |
Polyclonal | |
Unconjugated | |
SMG8 | |
1200011M11Rik; ABC2; Amplified in breast cancer gene 2 protein; BOS_18350; C17orf71; C19H17orf71; CG6729 gene product; FLJ10587; FLJ23205; LOC100353049; Protein SMG8; Protein smg-8 homolog; RGD1306862; Smg8; smg-8 homolog, nonsense mediated mRNA decay factor; smg-8 homolog, nonsense mediated mRNA decay factor (C. elegans); SMG8 nonsense mediated mRNA decay factor; SMG8 protein; SMG8, nonsense mediated mRNA decay factor | |
Rabbit | |
Protein A | |
RUO | |
55181 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q8ND04 | |
SMG8 | |
KLH conjugated synthetic peptide derived from human ABC2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction