Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABCA1 Antibody (5A1-1422.11) - BSA Free, Novus Biologicals™

Rat Monoclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NB400165
Description
ABCA1 Monoclonal specifically detects ABCA1 in Mouse samples. It is validated for Flow Cytometry, Western Blot (Negative).Specifications
| ABCA1 | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.1% Sodium Azide | |
| ABCA1 | |
| Mouse ABCA1-expressing HeLa stable transfectants. [UniProt# P41233] | |
| 0.1 mL | |
| ABC Transporters, Astrocyte Markers, Cancer, Cellular Markers, Cholesterol Metabolism, Glia Markers, Golgi Apparatus Markers, Lipid and Metabolism, Neuroscience, Signal Transduction | |
| 19 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Flow Cytometry, Western Blot | |
| 5A1-1422.11 | |
| Flow Cytometry 1:1000-1:3000, Western Blot (Negative) | |
| ABC-1, ABC1ATP-binding cassette transporter A1, ATP-binding cassette 1, ATP-binding cassette transporter 1, ATP-binding cassette, sub-family A (ABC1), member 1, CERP, Cholesterol efflux regulatory protein, EC 2.7.7.8, EC 2.8.1.8, EC 3.6.3, EC 3.6.3.41, FLJ14958, HDLDT1, membrane-bound, MGC164864, MGC165011, TGDATP-binding cassette sub-family A member 1 | |
| Rat | |
| Unpurified | |
| RUO | |
| Primary | |
| Mouse | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction