Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABCA1 Antibody (5A1-1422.11) - BSA Free, Novus Biologicals™

Rat Monoclonal Antibody has been used in 1 publication
$206.50 - $512.00
Specifications
| Antigen | ABCA1 |
|---|---|
| Clone | 5A1-1422.11 |
| Dilution | Flow Cytometry 1:1000-1:3000, Western Blot (Negative) |
| Applications | Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB400165SS
![]() |
Novus Biologicals
NB400165SS |
0.025 mL |
Each for $206.50
|
|
|||||
NB400165
![]() |
Novus Biologicals
NB400165 |
0.1 mL |
Each for $512.00
|
|
|||||
Description
ABCA1 Monoclonal specifically detects ABCA1 in Mouse samples. It is validated for Flow Cytometry, Western Blot (Negative).Specifications
| ABCA1 | |
| Flow Cytometry 1:1000-1:3000, Western Blot (Negative) | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Mouse | |
| ABC-1, ABC1ATP-binding cassette transporter A1, ATP-binding cassette 1, ATP-binding cassette transporter 1, ATP-binding cassette, sub-family A (ABC1), member 1, CERP, Cholesterol efflux regulatory protein, EC 2.7.7.8, EC 2.8.1.8, EC 3.6.3, EC 3.6.3.41, FLJ14958, HDLDT1, membrane-bound, MGC164864, MGC165011, TGDATP-binding cassette sub-family A member 1 | |
| ABCA1 | |
| IgG2a κ | |
| Unpurified |
| 5A1-1422.11 | |
| Flow Cytometry, Western Blot | |
| Unconjugated | |
| Rat | |
| ABC Transporters, Astrocyte Markers, Cancer, Cellular Markers, Cholesterol Metabolism, Glia Markers, Golgi Apparatus Markers, Lipid and Metabolism, Neuroscience, Signal Transduction | |
| Ascites with 0.1% Sodium Azide | |
| 19 | |
| Mouse ABCA1-expressing HeLa stable transfectants. [UniProt# P41233] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title