Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABCA1 Antibody (5A1-1422.11), DyLight 650, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB400165C
Description
ABCA1 Monoclonal specifically detects ABCA1 in Mouse samples. It is validated for Flow Cytometry.Specifications
ABCA1 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ABCA1 | |
Mouse ABCA1-expressing HeLa stable transfectants. [UniProt# P41233] | |
0.1 mL | |
ABC Transporters, Astrocyte Markers, Cancer, Cardiovascular Biology, Cellular Markers, Cholesterol Metabolism, Diabetes Research, Glia Markers, Golgi Apparatus Markers, Lipid and Metabolism, Neuroscience, Signal Transduction | |
Mouse | |
IgG2a κ |
Flow Cytometry | |
5A1-1422.11 | |
Flow Cytometry | |
ABC-1, ABC1ATP-binding cassette transporter A1, ATP-binding cassette 1, ATP-binding cassette transporter 1, ATP-binding cassette, sub-family A (ABC1), member 1, CERP, Cholesterol efflux regulatory protein, EC 2.7.7.8, EC 2.8.1.8, EC 3.6.3, EC 3.6.3.41, FLJ14958, HDLDT1, membrane-bound, MGC164864, MGC165011, TGDATP-binding cassette sub-family A member 1 | |
Rat | |
Protein A or G purified | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction