Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABCA1 Antibody, Allophycocyanin/Cy7, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB400105APCCY7
Description
ABCA1 Polyclonal specifically detects ABCA1 in Human, Mouse, Rat, Porcine, Canine, Chicken, Chinese Hamster, Equine, Hamster, Mustelid, Primate, Rabbit samples. It is validated for Flow Cytometry, Knockout Validated, Knockdown Validated.Specifications
| ABCA1 | |
| Polyclonal | |
| Flow Cytometry, Knockout Validated, Knockdown Validated | |
| ABC-1, ABC1ATP-binding cassette transporter A1, ATP-binding cassette 1, ATP-binding cassette transporter 1, ATP-binding cassette, sub-family A (ABC1), member 1, CERP, Cholesterol efflux regulatory protein, EC 2.7.7.8, EC 2.8.1.8, EC 3.6.3, EC 3.6.3.41, FLJ14958, HDLDT1, membrane-bound, MGC164864, MGC165011, TGDATP-binding cassette sub-family A member 1 | |
| Rabbit | |
| Affinity Purified | |
| ABC Transporters, Astrocyte Markers, Cancer, Cellular Markers, Cholesterol Metabolism, Glia Markers, Golgi Apparatus Markers, Lipid and Metabolism, Neuroscience, Signal Transduction | |
| 19.0 | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry, Immunoassay, KnockDown | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| ABCA1 | |
| Partial peptide sequence (within residues 1100-1300) of human ABCA1 [UniProt No. O95477]. Actual immunogen sequence is proprietary information. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Pig, Canine, Chicken, Chinese Hamster, Equine, Hamster, Mustelid, Primate, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction