Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABCA1 Antibody (HJ1), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. N102068APC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N102068APC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N102068APC7 Supplier Novus Biologicals Supplier No. NB1002068APCCY7
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ABCA1 Monoclonal specifically detects ABCA1 in Human, Mouse, Rat, Rabbit samples. It is validated for Flow Cytometry.

Specifications

Antigen ABCA1
Applications Flow Cytometry
Classification Monoclonal
Clone HJ1
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias ABC-1, ABC1ATP-binding cassette transporter A1, ATP-binding cassette 1, ATP-binding cassette transporter 1, ATP-binding cassette, sub-family A (ABC1), member 1, CERP, Cholesterol efflux regulatory protein, EC 2.7.7.8, EC 2.8.1.8, EC 3.6.3, EC 3.6.3.41, FLJ14958, HDLDT1, membrane-bound, MGC164864, MGC165011, TGDATP-binding cassette sub-family A member 1
Gene Symbols ABCA1
Host Species Mouse
Immunogen 50 kDa N-terminal extracellular loop of mouse ABCA1 [UniProt No. P41233]
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline ABC Transporters, Astrocyte Markers, Cancer, Cellular Markers, Cholesterol Metabolism, Glia Markers, Golgi Apparatus Markers, Lipid and Metabolism, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 19.0
Target Species Human, Mouse, Rat, Rabbit
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.