Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABCA1 Antibody (HJ1) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280554
Description
ABCA1 Monoclonal specifically detects ABCA1 in Human, Mouse, Rat, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
ABCA1 | |
Monoclonal | |
1 mg/mL | |
Western Blot : 1:2000, Flow Cytometry 1 ug per million cells. Use reported in scientific literature, Immunohistochemistry : 1:400, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 31061090), Immunohistochemistry-Paraffin : 1:400, CyTOF-ready | |
ABC-1, ABC1ATP-binding cassette transporter A1, ATP-binding cassette 1, ATP-binding cassette transporter 1, ATP-binding cassette, sub-family A (ABC1), member 1, CERP, Cholesterol efflux regulatory protein, EC 2.7.7.8, EC 2.8.1.8, EC 3.6.3, EC 3.6.3.41, FLJ14958, HDLDT1, membrane-bound, MGC164864, MGC165011, TGDATP-binding cassette sub-family A member 1 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, rat, rabbit and mouse. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG2b κ |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
HJ1 | |
Unconjugated | |
PBS | |
ABCA1 | |
50 kDa N-terminal extracellular loop of mouse ABCA1 [UniProt# P41233] | |
0.1 mL | |
ABC Transporters, Astrocyte Markers, Cancer, Cardiovascular Biology, Cellular Markers, Cholesterol Metabolism, Diabetes Research, Glia Markers, Golgi Apparatus Markers, Lipid and Metabolism, Neuroscience, Signal Transduction | |
19 | |
Human, Mouse, Rat, Rabbit | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction