Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABCA1 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB400105MFV450
Description
ABCA1 Polyclonal antibody specifically detects ABCA1 in Human, Mouse, Rat, Porcine, Canine, Chicken, Chinese Hamster, Equine, Hamster, Mustelid, Primate, Rabbit samples. It is validated for Flow CytometrySpecifications
ABCA1 | |
Polyclonal | |
Flow Cytometry | |
ABC-1, ABC1ATP-binding cassette transporter A1, ATP-binding cassette 1, ATP-binding cassette transporter 1, ATP-binding cassette, sub-family A (ABC1), member 1, CERP, Cholesterol efflux regulatory protein, EC 2.7.7.8, EC 2.8.1.8, EC 3.6.3, EC 3.6.3.41, FLJ14958, HDLDT1, membrane-bound, MGC164864, MGC165011, TGDATP-binding cassette sub-family A member 1 | |
Partial peptide sequence (within residues 1100-1300) of human ABCA1 Antibody [UniProt# O95477]. Actual immunogen sequence is proprietary information. | |
0.1 mL | |
ABC Transporters, Astrocyte Markers, Cancer, Cardiovascular Biology, Cellular Markers, Cholesterol Metabolism, Diabetes Research, Glia Markers, Golgi Apparatus Markers, Lipid and Metabolism, Neuroscience, Signal Transduction | |
19 | |
Store at 4C in the dark. | |
IgG |
Flow Cytometry | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Pig, Canine, Chicken, Chinese Hamster | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction