Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABCA1 Monoclonal Antibody (HJ1)
GREENER_CHOICE

Catalog No. PIMA516026
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA516026 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA516026 Supplier Invitrogen™ Supplier No. MA516026
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Prior to immunostaining paraffin tissues, antigen retrieval with sodium citrate buffer (pH 6.0) is recommended.

Mutations in the ABCA1 gene (ATP-binding cassette transporter 1) have been reported in Tangier disease (TD). TD is an autosomal recessive disorder characterized by an absence of plasma HDL, cholesterol ester depositing in the reticulo-endothelial system and deviations in cellular lipid trafficking. ABCA1 mediates the apo-A1 associated export of cholesterol and phospholipids from the cell. It is expressed on the plasma membrane and the Golgi complex, and is regulated by cholesterol flux. Regulation of the cholesterol flux between HDL and macrophages is competitive between ABCA1 and SR-BI.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCA1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone HJ1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene Abca1
Gene Accession No. O95477, P41233
Gene Alias ABC 1; ABC 1 antibody; ABC Transporter 1; Abc1; ABC-1; ABC1 antibody; Abca1; ATP binding cassette subfamily A member 1; ATP-binding cassette 1; ATP-binding cassette sub-family A member 1; ATP-binding cassette transporter 1; ATP-binding cassette transporter A1; ATP-binding cassette, subfamily A (ABC1), member 1; ATP-binding cassette, sub-family A (ABC1), member 1; ATP-binding cassette, sub-family A member 1; CERP; CERP antibody; Cholesterol efflux regulatory protein; HDLDT1; membrane-bound; phospholipid-transporting ATPase ABCA1; phospholipid-transporting ATPase ABCA1; ATP-binding cassette sub-family A member 1; TGD
Gene Symbols Abca1
Host Species Mouse
Immunogen 50 kDa N-terminal extracellular loop of mouse ABCA1.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11303, 19, 313210
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.