Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABCA1 Polyclonal Antibody, DyLight™ 488
GREENER_CHOICE

Catalog No. PIPA522908
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA522908 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA522908 Supplier Invitrogen™ Supplier No. PA522908
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ABCA1 Polyclonal Antibody for ChIP, ELISA, EMSA, Flow, ICC/IF, IHC (F), IHC (P), IP, ISH, Neu, Western Blot

Mutations in the ABCA1 gene (ATP-binding cassette transporter 1) have been reported in Tangier disease (TD). TD is an autosomal recessive disorder characterized by an absence of plasma HDL, cholesterol ester depositing in the reticulo-endothelial system and deviations in cellular lipid trafficking. ABCA1 mediates the apo-A1 associated export of cholesterol and phospholipids from the cell. It is expressed on the plasma membrane and the Golgi complex, and is regulated by cholesterol flux. Regulation of the cholesterol flux between HDL and macrophages is competitive between ABCA1 and SR-BI.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCA1
Applications ChIP Assay, ELISA, Flow Cytometry, Gel Shift, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, In Situ Hybridization (ISH), Neutralization, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.68 mg/mL
Conjugate DyLight 488
Formulation 50mM sodium borate with 0.05% sodium azide
Gene Abca1
Gene Accession No. O95477, P41233
Gene Alias ABC 1; ABC 1 antibody; ABC Transporter 1; Abc1; ABC-1; ABC1 antibody; Abca1; ATP binding cassette subfamily A member 1; ATP-binding cassette 1; ATP-binding cassette sub-family A member 1; ATP-binding cassette transporter 1; ATP-binding cassette transporter A1; ATP-binding cassette, subfamily A (ABC1), member 1; ATP-binding cassette, sub-family A (ABC1), member 1; ATP-binding cassette, sub-family A member 1; CERP; CERP antibody; Cholesterol efflux regulatory protein; HDLDT1; membrane-bound; phospholipid-transporting ATPase ABCA1; phospholipid-transporting ATPase ABCA1; ATP-binding cassette sub-family A member 1; TGD
Gene Symbols Abca1
Host Species Rabbit
Immunogen Partial peptide sequence (within residues 1100-1300) of human ABCA1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100054241, 100152112, 100356765, 100761226, 11303, 19, 313210, 373945, 481651
Target Species Canine, Chicken, Horse, Hamster, Human, Mustelid, Mouse, Monkey, Pig, Rabbit, Rat
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.