Learn More
Invitrogen™ ABCA1 Polyclonal Antibody, HRP

Description
ABCA1 Polyclonal Antibody for Western Blot, IHC (P), IHC (F), ICC/IF, Flow, ELISA, IP, ChIP, Neu, GS, ISH
Specifications
Specifications
| Antigen | ABCA1 |
| Applications | ChIP Assay, ELISA, Flow Cytometry, Gel Shift, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, In Situ Hybridization (ISH), Neutralization, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.66 mg/mL |
| Conjugate | HRP |
| Formulation | PBS with no preservative |
| Gene | Abca1 |
| Gene Accession No. | O95477, P41233 |
| Gene Alias | ABC 1; ABC 1 antibody; ABC Transporter 1; Abc1; ABC-1; ABC1 antibody; Abca1; ATP binding cassette subfamily A member 1; ATP-binding cassette 1; ATP-binding cassette sub-family A member 1; ATP-binding cassette transporter 1; ATP-binding cassette transporter A1; ATP-binding cassette, subfamily A (ABC1), member 1; ATP-binding cassette, sub-family A (ABC1), member 1; ATP-binding cassette, sub-family A member 1; CERP; CERP antibody; Cholesterol efflux regulatory protein; HDLDT1; membrane-bound; phospholipid-transporting ATPase ABCA1; phospholipid-transporting ATPase ABCA1; ATP-binding cassette sub-family A member 1; TGD |
| Gene Symbols | Abca1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.