Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABCA10 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305791JF646
Description
ABCA10 Polyclonal specifically detects ABCA10 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ABCA10 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
ATP-binding cassette A10, ATP-binding cassette sub-family A member 10, ATP-binding cassette, sub-family A (ABC1), member 10, EC 3.6.3, EC 3.6.3.41, EST698739 | |
Produced in rabbits immunized with E. coli-derived Human ABCA10 fragment. | |
0.1 mL | |
Protein Kinase | |
10349 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction