Learn More
ABCA10 Monoclonal Antibody (8F4)

Description
ABCA10 Monoclonal Antibody for Western Blot, ELISA
Specifications
Specifications
| Antigen | ABCA10 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 8F4 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | ABCA10 |
| Gene Accession No. | Q8WWZ4 |
| Gene Alias | ABCA10; ATP binding cassette subfamily A member 10; ATP-binding cassette A10; ATP-binding cassette sub-family A member 10; ATP-binding cassette, sub-family A (ABC1), member 10; EST698739; FKHL6; FREAC2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.