Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABCA12 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA5122304
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5122304 20 μL
PIPA5122303 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5122304 Supplier Invitrogen™ Supplier No. PA5122304
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intracellular membranes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCA12
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene ABCA12
Gene Accession No. Q86UK0
Gene Alias 4832428G11Rik; 4833417A11Rik; ABC12; ABCA12; ARCI4A; ARCI4B; ATP binding cassette subfamily A member 12; ATP-binding cassette 12; ATP-binding cassette sub-family A member 12; ATP-binding cassette transporter 12; ATP-binding cassette, subfamily A (ABC1), member 12; ATP-binding cassette, sub-family A (ABC1), member 12; ATP-binding cassette, sub-family A, member 12; DKFZp434G232; FLJ41584; ICR2B; LI2
Gene Symbols ABCA12
Host Species Rabbit
Immunogen Recombinant ABCA12 (Val1346~Thr1577) with two N-terminal Tags, His-tag and T7-tag expressed in E.coli.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26154
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.