Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABCA2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | ABCA2 |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
ABCA2 Polyclonal specifically detects ABCA2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ABCA2 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
Q9BZC7 | |
20 | |
This antibody was developed against a recombinant protein corresponding to the amino acids: PTISVKEAFYTAAPLTSAGILPVMQSLCPDGQRDEFGFLQYANSTVTQLLERLDRVVEEGNLFDPARPSLGSELEALRQHLEALSAGPGTSGSHLDRSTVSSFSLDSV | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Polyclonal | |
Rabbit | |
ABC Transporters, Lipid and Metabolism | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
ABC2ATP-binding cassette, sub-family A, member 2, ATP-binding cassette 2, ATP-binding cassette sub-family A member 2, ATP-binding cassette transporter 2, ATP-binding cassette, sub-family A (ABC1), member 2, EC 3.6.3, EC 3.6.3.41, KIAA1062, MGC129761 | |
ABCA2 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title