Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ABCA2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560157
Description
Immunogen sequence: PTISVKEAFY TAAPLTSAGI LPVMQSLCPD GQRDEFGFLQ YANSTVTQLL ERLDRVVEEG NLFDPARPSL GSELEALRQH LEALSAGPGT SGSHLDRSTV SSFSLDSV Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 89%.
Probable transporter, its natural substrate has not been found yet. May have a role in macrophage lipid metabolism and neural development.
Specifications
ABCA2 | |
Polyclonal | |
Unconjugated | |
Abca2 | |
Abc2; ABCA2; abca2 {ECO:0000312; AI413825; ATP binding cassette subfamily A member 2; ATP-binding cassette 2; ATP-binding cassette sub-family A member 2; ATP-binding cassette transporter 2; ATP-binding cassette, subfamily A (ABC1), member 2; ATP-binding cassette, sub-family A (ABC1), member 2; ATP-binding cassette, sub-family A, member 2; D2H0S1474E; KIAA1062; mKIAA1062; RGD:620238} | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BZC7 | |
Abca2 | |
Recombinant protein corresponding to Human ABCA2. Recombinant protein control fragment (Product #RP-92911). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction