Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABCA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120484
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120484 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120484 Supplier Invitrogen™ Supplier No. PA5120484
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HeLa, Rat brain. The target is usually found in the following locations: Endosome membrane, Lysosome membrane, Multi-pass membrane protein.

Probable transporter, its natural substrate has not been found yet. May have a role in macrophage lipid metabolism and neural development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCA2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Abca2
Gene Accession No. P41234, Q9BZC7, Q9ESR9
Gene Alias Abc2; ABCA2; abca2 {ECO:0000312; AI413825; ATP binding cassette subfamily A member 2; ATP-binding cassette 2; ATP-binding cassette sub-family A member 2; ATP-binding cassette transporter 2; ATP-binding cassette, subfamily A (ABC1), member 2; ATP-binding cassette, sub-family A (ABC1), member 2; ATP-binding cassette, sub-family A, member 2; D2H0S1474E; KIAA1062; mKIAA1062; RGD:620238}
Gene Symbols Abca2
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11305, 20, 79248
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.