Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABCA3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5118968
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118968 200 μL
PIPA5122302 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5118968 Supplier Invitrogen™ Supplier No. PA5118968
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ABCA3 Polyclonal Antibody for Western Blot

Plays an important role in the formation of pulmonary surfactant, probably by transporting lipids such as cholesterol.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCA3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene ABCA3
Gene Accession No. Q99758
Gene Alias 1810036E22Rik; ABC transporter 3; ABC3; ABCA3; ABC-C; ABC-C transporter; ATP binding cassette subfamily A member 3; ATP-binding cassette 3; ATP-binding cassette sub-family A member 3; ATP-binding cassette transporter 3; ATP-binding cassette, subfamily A (ABC1), member 3; ATP-binding cassette, sub-family A (ABC1), member 3; EST111653; LBM180; SMDP3
Gene Symbols ABCA3
Host Species Rabbit
Immunogen Recombinant protein ATP Binding Cassette Transporter A3. The antigen corresponds to amino acid range 1358-1635 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21, 302973
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.