Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABCA4 Antibody (3F4), Novus Biologicals™
SDP

Catalog No. NBP130032 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP130032 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP130032 Supplier R&D Systems Supplier No. NBP130032

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody has been used in 4 publications

ABCA4 Monoclonal specifically detects ABCA4 in Bovine, Canine, Feline, Human, Mouse, Xenopus species, Xenopus species (African Clawed Frog) samples. It is validated for Gene Knock-Out, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot.

Specifications

Antigen ABCA4
Applications Gene Knock-Out, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone 3F4
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen 1:10 - 1:500, Knockout Validated
Gene Alias ABC10, ABCRRMP, ARMD2DKFZp781N1972, ATP binding cassette transporter, ATP-binding cassette sub-family A member 4, ATP-binding cassette transporter, retinal-specific, ATP-binding cassette, sub-family A (ABC1), member 4, ATP-binding transporter, retina-specific, CORD3, EC 3.6.3, FFMFLJ17534, photoreceptor rim protein, retinal-specific ATP-binding cassette transporter, retina-specific ABC transporter, RIM ABC transporter, RIM protein, RmP, RP19, Stargardt disease protein, STGD, STGD1
Gene Symbols ABCA4
Host Species Mouse
Immunogen Partially purified bovine 220-kDa disc rim protein. Accession # F1MWM0
Molecular Weight of Antigen 220 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline ABC Transporters, Lipid and Metabolism, Vision
Primary or Secondary Primary
Gene ID (Entrez) 24
Target Species Bovine, Canine, Feline, Human, Mouse, Xenopus species, Xenopus species
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.